U-Channel Sign Posts, Brady®
Supplier: Brady Worldwide
95047 97204 97205 97207 97208 97209 97210
CA89007-918EA 93.7 CAD
CA89007-918 CA89007-938 CA89007-940 CA89007-944 CA89007-946 CA89007-948 CA89007-950
U-Channel Sign Posts, Brady®
Signs
Sign posts are ideal for mounting and installing traffic, utility, and industrial signs
Standard U-channel posts have a baked-green enamel finish on hot-rolled, high-tensile strength steel, allowing for years of maintenance-free performance. These sign posts weigh 0.9kg/linear meter (2lbs./linear foot) and have 9.5mm (3/8") holes, 2.5cm (1") on center. To fasten signs onto standard U-channel posts, use the vandal-proof Tufnut fastener. The Tufnut fastener is an 8mm (5/16") plated steel nut that secures tightly to prevent sign theft. Complete Tufnut set includes 2 bolts, 2 washers, and 3More Product Information