Small Universal Valve Lockout Base Clamping Unit, Brady Worldwide®
Supplier: Brady Worldwide
50924
CA89007-222EA 66.6 CAD
CA89007-222
Small Universal Valve Lockout Base Clamping Unit, Brady Worldwide®
Clamps General Purpose Clamps
The universal valve lockouts are effective for all types of standard valves.
  • Versatile
  • Rugged
  • Easy-to-use
  • Lightweight
They can also be used to lock out large levers, T-handles, and hard to secure mechanical devices. New open-ended clamp fits over closed rings and wide handles. Constructed of industrial-grade steel and nylon for extra impact and chemical resistance.
Ordering information: Base unit only (cable and blocking arm sold separately).