BX™ Series Protective Eyewear, 3M™
Supplier: 3M
7000052795 7000052796
CA89006-497CS 221.06 CAD
CA89006-497 CA89006-545
BX™ Series Protective Eyewear, 3M™
Safety Glasses
BX™ series protective eyewear features a soft fully adjustable nose bridge, lens angle adjustment, and adjustable length temples for extended wear.
  • Nose bridge is cushioned for softness and comfort
  • Sleek, contoured design offers an attractive appearance along with functional eye protection
  • Impact-resistant polycarbonate lenses absorb 99.9% UV rays
The ability to customize the safety glasses to the wearer makes them more pleasant to use over extended time periods.
Eye protection is important to safeguard the user's eyes and maintain healthy vision.More Product Information