Hydrogen peroxide 30 - 32%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®

Supplier: VWR International
Danger

CA87003-224 87003-224 76186-226
CA87003-224EA 2524.56 CAD
CA87003-224 87003-224 76186-226
Hydrogen peroxide 30 - 32%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Hydrogen peroxide

Ultra high purity acid for quantitative trace metal analysis at the parts-per-trillion (ppt) level.

Formula: H₂O₂
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Assay 30 - 32 %
Aluminum (Al) max. 50 ppt
Antimony (Sb) max. 10 ppt
Arsenic (As) max. 100 ppt
Barium (Ba) max. 10 ppt
Beryllium (Be) max. 10 ppt
Bismuth (Bi) max. 10 ppt
Boron (B) max. 100 ppt
Cadmium (Cd) max. 10 ppt
Calcium (Ca) max. 100 ppt
Cerium (Ce) max. 1 ppt
Cesium (Cs) max. 1 ppt
Chromium (Cr) max. 10 ppt
Cobalt (Co) max. 10 ppt
Copper (Cu) max. 10 ppt
Dysprosium (Dy) max. 1 ppt
Erbium (Er) max. 1 ppt
Europium (Eu) max. 1 ppt
Gadolinium (Gd) max. 1 ppt
Gallium (Ga) max. 10 ppt
Germanium (Ge) max. 10 ppt
Gold (Au) max. 10 ppt
Hafnium (Hf) max. 1 ppt
Holmium (Ho) max. 1 ppt
Indium (In) max. 1 ppt
Iron (Fe) max. 20 ppt
Lanthanum (La) max. 1 ppt
Lead (Pb) max. 10 ppt
Lithium (Li) max. 10 ppt
Lutetium (Lu) max. 1 ppt
Magnesium (Mg) max. 20 ppt
Manganese (Mn) max. 10 ppt
Mercury (Hg) max. 50 ppt
Molybdenum (Mo) max. 10 ppt
Neodymium (Nd) max. 1 ppt
Nickel (Ni) max. 20 ppt
Niobium (Nb) max. 10 ppt
Palladium (Pd) max. 10 ppt
Potassium (K) max. 20 ppt
Praseodymium (Pr) max. 1 ppt
Rhenium (Re) max. 10 ppt
Rhodium (Rh) max. 10 ppt
Rubidium (Rb) max. 10 ppt
Ruthenium (Ru) max. 10 ppt
Samarium (Sm) max. 1 ppt
Scandium (Sc) max. 10 ppt
Selenium (Se) max. 100 ppt
Silver (Ag) max. 10 ppt
Sodium (Na) max. 50 ppt
Strontium (Sr) max. 10 ppt
Tantalum (Ta) max. 10 ppt
Tellurium (Te) max. 1 ppt
Terbium (Tb) max. 1 ppt
Thallium (Tl) max. 1 ppt
Thorium (Th) max. 1 ppt
Thulium (Tm) max. 1 ppt
Tin (Sn) max. 50 ppt
Titanium (Ti) max. 20 ppt
Tungsten (W) max. 20 ppt
Uranium (U) max. 1 ppt
Vanadium (V) max. 10 ppt
Ytterbium (Yb) max. 1 ppt
Yttrium (Y) max. 1 ppt
Zinc (Zn) max. 50 ppt
Zirconium (Zr) max. 10 ppt

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR