EasyDip™ Slide Staining System, Simport Scientific
Supplier: Simport Scientific
M900-12AS M900-12B M900-12G M900-12P M900-12W M900-12Y M905-12DGY M900-12B M906-12AS
CA-87000-130CS 211.35 CAD
CA-87000-130 CA87000-120 CA87000-122 CA87000-124 CA87000-126 CA87000-128 CA87000-132 87000-120 CA10154-052
EasyDip™ Slide Staining System, Simport Scientific
Staining Boxes
The slide staining system is made of acetal plastic.
  • Jars can be loosely joined to each other to keep the same order when moved around
  • Wide stable base for stability while inside is recessed to reduce reagent volume
  • Available in five colours to aid identification of contents or applications
The staining jar is made of resistant acetal plastic, and will not break like most glass jars do. It will resist most staining agents including alcohol and xylene (but not phenol, iodine, or ferric chloride). The wide stable base offers greater stability while theMore Product Information