Stain Kits and Replacement Components, Epredia
Supplier: Epredia
87101 87015 88001 88003 88117 87016 88002 87003 88004 88005 88006 87017 88007 88008 88009 88000 88011 87018 88012 88101 88102 88103 88104 87006 88013 88014 88015 87007 88017 88018 87008 88023 88024 88025 88026 88027 88036 87009 88033 88034 88035 87019 88019 88020 88022 88028 88029 88038 88039 87012 88037 87020 88030 87021 88031
16002-128EA 1180.88 CAD
16002-128 CA84000-200 CA84000-202 CA84000-204 CA84000-206 CA84000-208 CA84000-210 CA84000-212 CA84000-214 CA84000-216 CA84000-218 CA84000-220 CA84000-222 CA84000-224 CA84000-226 CA84000-228 CA84000-230 CA84000-232 CA84000-234 CA84000-236 CA84000-238 CA84000-240 CA84000-242 CA84000-244 CA84000-246 CA84000-248 CA84000-250 CA84000-252 CA84000-256 CA84000-258 CA84000-260 CA84000-262 CA84000-264 CA84000-266 CA84000-268 CA84000-270 CA84000-272 CA84000-274 CA84000-280 CA84000-282 CA84000-284 CA84000-286 CA84000-288 CA84000-290 CA84000-292 CA84000-294 CA84000-296 CA84000-298 CA84000-300 CA84000-304 CA84000-306 CA84000-308 CA84000-310 CA84000-312 CA84000-314
Stain Kits and Replacement Components, Epredia
Stains Histology Stains
Epredia™ stain kits and replacement components contain all the necessities of the staining procedure.
  • Save time
  • Components available separately
  • All the necessary stains, solutions and components
Acid Fast Bacillus, blue secondary stain kits stain bright red with a pale blue background. Acid Fast Bacillus, green secondary stain kit stains dark red with a pale green background. Demonstrate amyloid with Epredia™ Amyloid Stain. Amyloid stain kit stains orange to red.
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Elastic Stain. Elastic stain kits stain blue-black to black for elastic tissue, red for collagen with a yellow background.
Save time with Epredia™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome. Gomori Trichrome (Blue Collagen) stain kit stains red for Muscle tissue and cytoplasm, blue for collagen and blue-black for nuclei. Epredia™ Gomori Trichrome (Green Collagen is a one-step trichrome stain that provides distinctive red muscle fibers with green collagen staining.
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Iron Stain. Iron stain kits stain blue iron, pale pink cytoplasm and pink-red nuclei.
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid), a uniquely modified silver stain formulation that utilizes methenamine/borax capsules to make working silver solution. Modified Grocott’s Methenamine kit stains black with a green background.
Identify primary tumor sites with the aid of Epredia™ Mucicarmine. Mucicarmine stain kit stains dark rose for mucin and black for nuclei with a yellow background.
Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Periodic Acid-Schiff (PAS) Solution. Periodic Acid Schiff kit stains rose to magenta with blue nuclei.
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Steiner-Steiner Silver. Steiner-Steiner silver kits stain black with a yellow to brown background.
Ordering information: All kits come with all stains and solutions necessary to complete the staining procedure. Components of kits are also available separately.