Papanicolaou's Orange G solution (OG 6)
Supplier: RICHARD-ALLAN SCIENTIFIC CA
75204 75211 75225
CA84000-134CS 679.73 CAD
CA84000-134 CA84000-136 CA84000-138
Papanicolaou's Orange G solution (OG 6)
Papanicolaou's Orange G solution (OG 6)
Stain gynecological specimens with Epredia™ Papanicolaou OG-6 Stain. OG-6 Stain is a traditional formulation that, when used with various EA products, will provide familiar staining results for gynecological and non-gynecological specimens.
IVDLogo
Order Now
SPECIFICATIONS
Environmentally Preferable