Single Tissue Multiple Species Blot, G-Biosciences
Supplier: G-Biosciences
TB28 TB29 TB30 TB31 TB32 TB33 TB34 TB35 TB36
CA82023-184EA 2188.17 CAD
CA82023-184 CA82023-186 CA82023-188 CA82023-190 CA82023-192 CA82023-194 CA82023-196 CA82023-198 CA82023-200
Single Tissue Multiple Species Blot, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, single tissue, multi-species blots (human, mouse and rat).
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department forMore Product Information