Rat Single Tissue Single Species Blot, G-Biosciences
Supplier: G-Biosciences
TB03 TB06 TB09 TB12 TB15 TB18 TB21 TB24 TB27
CA82023-134EA 1479.2 CAD
CA82023-134 CA82023-140 CA82023-146 CA82023-152 CA82023-158 CA82023-164 CA82023-170 CA82023-176 CA82023-182
Rat Single Tissue Single Species Blot, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, single tissue (normal) rat blots.
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information onMore Product Information