Ammonium acetate solution 5M

Supplier: G-Biosciences

Synonyms: Acetic acid ammonium salt

R012
CA82023-100EA 170.8 CAD
CA82023-100
Ammonium acetate solution 5M
Ammonium acetate
Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Flash Pt: 136
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
Merck Index: 13,00495

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR