SpinOUT™ Protein Desalting and Buffer Exchange Columns, G-Biosciences
Supplier: G-Biosciences
786-170 786-171 786-172 786-173 786-174 786-175 786-703 786-704 786-705 786-706 786-707 786-708
CA82022-580PK 314.53 CAD
CA82022-580 CA82022-582 CA82022-584 CA82022-586 CA82022-588 CA82022-590 CA89167-920 CA89167-922 CA89167-924 CA89167-926 CA89167-928 CA89167-930
SpinOUT™ Protein Desalting and Buffer Exchange Columns, G-Biosciences
Chromatography Columns
The SpinOUT™ GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of protein solutions ranging from 5µL through to 4mL sample volumes
The SpinOUT™ columns are simply to use as the protein sample is applied, and then centrifuged to recover protein, with the column retaining >95% of the salts and small molecules (<1,000Da for GT-600 and <1,500 for GT-1200). Spin-OUT™ GT-600 is for the purification of proteins > 6kDa and nucleic acids larger than 10bp. Spin-OUT™ GT-1200 is for the purification of proteins > 30kDa and removal ofMore Product Information