Mammalian GenLysate™ Pig Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
ML-07 SLP-081 SLP-082 SLP-083 SLP-084 SLP-085 SLP-086 SLP-087 ML-10
CA82021-684EA 582.14 CAD
CA82021-684 CA82022-196 CA82022-200 CA82022-204 CA82022-208 CA82022-212 CA82022-216 CA82022-220 CA95029-320
Mammalian GenLysate™ Pig Total Protein Lysates, G-Biosciences
Cells
G-Biosciences' GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest. To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail. Each lysate lot is tested by electrophoresis and Western blot analysis.
G-Biosciences' GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes
G-Biosciences' GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris·HCl and 2% SDS with G-Biosciences' ProteaseARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.