Mammalian GenLysate™ Mouse Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
CFM-011 CFM-012 CFM-013 CFM-014 CFM-015 NLM-01 NLM-02 NLM-03 NLM-04 NLM-05 NLM-06 NLM-07 NLM-08 NLM-09 SLM-021 SLM-0211 SLM-0212 SLM-022 SLM-023 SLM-024 SLM-025 SLM-026 SLM-027 SLM-028 SLM-029 SLM-041 SLM-042 SLM-0210
CA82021-614EA 660.84 CAD
CA82021-614 CA82021-616 CA82021-618 CA82021-620 CA82021-622 CA82021-808 CA82021-812 CA82021-816 CA82021-820 CA82021-824 CA82021-828 CA82021-832 CA82021-836 CA82021-840 CA82022-140 CA82022-144 CA82022-148 CA82022-154 CA82022-158 CA82022-162 CA82022-166 CA82022-170 CA82022-174 CA82022-178 CA82022-182 CA82022-186 CA82022-190 CA95029-334
Mammalian GenLysate™ Mouse Total Protein Lysates, G-Biosciences
Cells
G-Biosciences' GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest. To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail. Each lysate lot is tested by electrophoresis and Western blot analysis.
G-Biosciences' GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes
G-Biosciences' GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris·HCl and 2% SDS with G-Biosciences' ProteaseARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.