Mammalian GenLysate™ Human Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
NLH-01 NLH-02 NLH-03 NLH-04 NLH-05 NLH-06 NLH-08 NLH-09 NLH-11 NLH-12 NLH-13 NLH-14 NLH-15 NLH-16 NLH-17 NLH-18 NLH-19 NLH-20 NLH-21 SLH-021 SLH-0210 SLH-0211 SLH-0212 SLH-0213 SLH-023 SLH-024 SLH-025 SLH-026 SLH-027 SLH-028 SLH-029 SLH-081 SLH-082 SLH-088 SLH-089 NLH-22 SLH-087 SLH-090
CA82021-728EA 682.77 CAD
CA82021-728 CA82021-732 CA82021-736 CA82021-740 CA82021-744 CA82021-748 CA82021-752 CA82021-756 CA82021-760 CA82021-764 CA82021-768 CA82021-772 CA82021-776 CA82021-780 CA82021-784 CA82021-788 CA82021-792 CA82021-796 CA82021-800 CA82021-804 CA82022-074 CA82022-076 CA82022-080 CA82022-084 CA82022-088 CA82022-096 CA82022-100 CA82022-104 CA82022-108 CA82022-112 CA82022-116 CA82022-120 CA82022-124 CA82022-128 CA82022-132 CA82022-136 CA95029-322 CA95029-330 CA95029-332
Mammalian GenLysate™ Human Total Protein Lysates, G-Biosciences
Cells
G-Biosciences GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes.
  • Offered as a 150 µg lyophilized protein per vial
  • Ready for use
  • Wide variety of species and tissues
G-Biosciences GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris HCl and 2% SDS with G-Biosciences Protease ARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.
G-Biosciences GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail.
Order Now
SPECIFICATIONS
Environmentally Preferable