Column-PROTEIN-Concentrate™ Concentration Kit, G-Biosciences
Supplier: G-Biosciences
786-126
CA82021-284EA 735.21 CAD
CA82021-284
Column-PROTEIN-Concentrate™ Concentration Kit, G-Biosciences
Protein Extraction Protein Extraction Kits
The Column-PROTEIN-Concentrate™ kit has been specifically developed for concentration of those proteins that cannot be concentrated either by precipitation or other techniques
The immobilized protein is spin-eluted in a small volume of specifically formulated elution buffer, giving several fold effective concentration. The method is gentle and protects protein from denaturation during the concentration process. The Column-PROTEIN-Concentrate™ kit maintains and protects the biological activity of proteins and allows for ~100% recovery.
This kit is suitable for concentration of a total of 4mg protein in either single or multiple procedures. For concentration of larger than 5mg protein, a custom made kit may be ordered. The kit is supplied with a proprietary spin column; the column may be regenerated and reused once.
The kit is based on a proprietary Protein Binding Resin that binds and immobilizes any protein in a low salt buffer between pH 2-12. The binding capacity is ~0.5mg protein/ml Protein Binding Resin.
Ordering information: Accessories not included.