KLEENGUARD® A40 Liquid and Particle Protection Shoe Covers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Microflex
44490 44492 44494
CA82007-666CS 401.17 CAD
CA82007-666 CA82007-668 CA82007-670
KLEENGUARD® A40 Liquid and Particle Protection Shoe Covers, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
Shoe covers provide an excellent liquid and particle barrier. They are designed to be used in applications where Tyvek® material is typically worn. KLEENGUARD® A40 is a lamination of microporous film and a spunbond nonwoven material. KLEENGUARD® A40 laminate has a dry particle holdout of 100% between 0.3–0.5µm.
  • Used in applications where Tyvek® material is typically worn
  • Lamination of microporous film and spunbond nonwoven material
  • Dry particle holdout of 100% between 0.3–0.5µm
Shoe covers have elastic at the tops
Meet ANSI/ISEA 101-1996.