Benstat™ Divider Tote Boxes, Metro™
Supplier: Metropolitan Wire
TB91060BAS TB92035BAS TB92050BAS TB92080BAS TB93030BAS TB93050BAS TB93060BAS TB93080BAS TB93120BAS
CA82002-522EA 75.6 CAD
CA82002-522 CA82002-525 CA82002-526 CA82002-530 CA82002-532 CA82002-534 CA82002-537 CA82002-538 CA82003-786
Benstat™ Divider Tote Boxes, Metro™
Boxes ESD-Safe Boxes
Tote boxes offer safe and efficient storage of items
Boxes, dividers, covers, and card holders are made from Benstat™, a static-dissipative polypropylene-based material with a surface resistivity of >105ohms/sq. and <1012ohms/sq. All Benstat™ products are blue.
Insert style covers fit inside the rim of the tote box and feature a finger hole that enables easy removal. Card holders are also available. Choose from holders made from Benstat™ or vinyl.
Boxes are durable, stackable, and feature high impact strength. There is a smooth area on each side of the box that is a convenientMore Product Information