Anti-c-K-Ras (234-4.2) Mouse Monoclonal Antibody

Supplier: MilliporeSigma
OP24-100UG
CA80504-820EA 1148.57 CAD
CA80504-820
Anti-c-K-Ras (234-4.2) Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-c-K-Ras (Ab-1) (234-4.2) Reacts with Human, Mouse, Rat

Type: Primary
Antigen: c-K-Ras (234-4.2)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR