Anti-COX2 Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
236002-50UG
CA80501-504EA 2308.58 CAD
CA80501-504
Anti-COX2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COX2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Sheep, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR