DL-α-Tocopherol, Millipore®

Supplier: MilliporeSigma

613420-50GM
CA80109-028EA 225.05 CAD
CA80109-028
DL-α-Tocopherol, Millipore®
DL-α-Tocopherol

A physiological membrane-bound antioxidant that protects cell membrane lipids from oxidative damage by free radicals.

Formula: C₂₉H₅₀O₂
MDL Number: MFCD00072051
CAS Number: 10191-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR