D-(-)-Mannitol

Supplier: MilliporeSigma

443907-100GM 443907-500GM
CA80108-990EA 178.73 CAD
CA80108-990 CA80108-992
D-(-)-Mannitol
D(-)-Mannitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MDL Number: MFCD00064287
CAS Number: 69-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR