Thermal Transfer Printable Labels, Brady
Supplier: Brady Worldwide
62354 62355 62359 62363 62365 62366 62371 62379
CA80090-256EA 225.29 CAD
CA80090-256 CA80090-258 CA80090-260 CA80090-262 CA80090-264 CA80090-266 CA80090-268 CA80090-270
Thermal Transfer Printable Labels, Brady
Labels
Printable, self-laminating polyester labels for thermal transfer printers.
  • Good print smudge resistance
  • Solvent resistance
  • Good high and low temperature performance
B-461 performs well in common laboratory environments such as liquid nitrogen and autoclave applications. Used for laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides. It can be printed with Series R6200 ribbon.
The B-461 self laminating label is for 1 to 2 ml cryo vials-short tail, slit in liner for easy removal. It can be offered in a self-More Product Information