X-plore® Air-Purifying Respirators, Draeger
Supplier: Draeger
R55270 R55331 R55330 R55332 R55351 R55350 R55352 6738024 6738025 6738027 6738034 6738035 6738037 6738020 6738021 6738350 6738351 6738361 6738392 6738816 6738817 6740830
CA80082-476EA 983.29 CAD
CA80082-476 CA80082-478 CA80082-480 CA80082-482 CA80082-484 CA80082-486 CA80082-488 CA80082-490 CA80082-492 CA80082-496 CA80082-498 CA80082-500 CA80082-504 CA80082-506 CA80082-508 76639-005 76638-977 76638-983 76638-979 76638-989 76639-009 76638-999
X-plore® Air-Purifying Respirators, Draeger
Respirators Reusable Respirators Half Face Respirators
Respirators provide comfortable protection during industrial tasks, such as welding, painting, or chemical handling.
  • All masks use a two-point bayonet connection for safe and easy filter attachment
The 5500 series full-face masks offer complete respiratory and eye protection
The 3500 series reusable half-masks employ unique Draeger-Flex™ hypoallergenic material that has a soft and comfortable seal without skin irritation. The FlexiFit® harness system is extremely comfortable even under a hard hat. These compact, low-profile masks can be easily wornMore Product Information