Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: Eppendorf
022431102 022431064 022431081 0030108302 0030108302 0030122216 0030122240 0030122356 0030108531 0030122283
CA80077-226PK 59.87 CAD
CA80077-226 CA80077-228 CA80077-232 89429-314 CA89429-314 76289-496 76289-498 76313-966 CA76683-144 CA76683-150
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
Engineered to minimize sample binding and elevate recovery, Eppendorf® Protein LoBind® tubes safeguard valuable proteins during processing. The cutting-edge LoBind® technology ensures optimal yields for Western blotting, mass spectrometry, and protein purification applications.
  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors
  • Available in tube, microplate, and deep-well plate formats for convenient
More Product Information