DuPont™ Tyvek® IsoClean® Hoods with Eyes Only Opening
Supplier: DuPont
IC666BWHMD01000C IC666BWHLG01000C IC666BWHMD0100CS IC666BWHLG010000 IC666BWHMD010000
CA80076-816CS 3267.88 CAD
CA80076-816 CA80076-818 CA80076-864 CA89129-688 CA89129-690
DuPont™ Tyvek® IsoClean® Hoods with Eyes Only Opening
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Bound seams
  • Bound opening for eyes
  • Bound bottom
  • Snaps for adjustable fit
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Bound hood opening ensures lower particle shedding. Eyes-only opening is 9" wide. Snaps allow for an adjustable fit.