ULTROL® Grade, Tris Base Buffer, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
648311-5KG
CA80058-772EA 1011.33 CAD
CA80058-772
ULTROL® Grade, Tris Base Buffer, Calbiochem®, MilliporeSigma
Buffers Biological Buffers
Tris Base buffer is useful in the pH range of 10.3 to 11.3 and has a pKa of 8.1 at 25 °C.
Has an absorbance <0.05 (1.0 M, H₂O, 260 nm).