Sodium salicylate ≥98%, Calbiochem®

Supplier: MilliporeSigma
Warning

Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt

567630-5GM
CA80058-660EA 156.89 CAD
CA80058-660
Sodium salicylate ≥98%, Calbiochem®
Sodium salicylate

A non-steroidal anti-inflammatory drug that interferes with TNF-induced NF-[kappa]B activation by inhibiting phosphorylation and subsequent degradation of the I[kappa]B-[alpha] protein. Selectively inhibits TNF-induced activation of p42 (ERK2) and p44 (ERK1) MAP kinases. Also inhibits TNF-induced activation of JNK. Induces apoptosis via p38 kinase activation. Inhibits COX-2 and inducible NOS (iNOS) transcription independently of NF-[kappa]B activation.

Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Melting Pt: 200
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 12,08819

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR