Disodium hydrogen phosphate

Supplier: MilliporeSigma

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

567550-500GM
CA80058-656EA 234.89 CAD
CA80058-656
Disodium hydrogen phosphate
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250
MDL Number: MFCD00003496
CAS Number: 7558-79-4
Merck Index: 12,08805

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR