N-Octyl-β-D-glucopyranoside, ULTROL®

Supplier: MilliporeSigma

494460-1GM 494460-250MG 494460-5GM
CA80058-522EA 728.88 CAD
CA80058-522 CA80058-524 CA80058-526
N-Octyl-β-D-glucopyranoside, ULTROL®
N-Octyl-β-D-glucopyranoside
Formula: C₁₄H₂₈O₆
MDL Number: MFCD00063288
CAS Number: 29836-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR