N-Octyl-β-D-glucopyranoside

Supplier: MilliporeSigma

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

494459-1GM 494459-25GM 494459-5GM
CA80058-514EA 244.2 CAD
CA80058-514 CA80058-516 CA80058-520
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Melting Pt: 110
MDL Number: MFCD00063288
CAS Number: 29836-26-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR