TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid), ULTROL®

Supplier: MilliporeSigma

Synonyms: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulfonic acid, 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulfonic acid

394675-1KG
CA80058-262EA 1195.67 CAD
CA80058-262
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid), ULTROL®
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Formula: C₇H₁₇NO₆S
MW: 243.28 g/mol
MDL Number: MFCD00007538
CAS Number: 29915-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR