Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside

Supplier: MilliporeSigma

373272-5GM
CA80058-178EA 787.21 CAD
CA80058-178
Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside
Methyl-6-O-(N-heptylcarbamoyl)-alpha-D-glucopyranoside
Formula: C₁₅H₂₉NO₇
MDL Number: MFCD00077425
CAS Number: 115457-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR