L(-)-Glutathione (oxidised form) ≥95% (by HPLC)

Supplier: MilliporeSigma

3542-5GM
CA80056-570EA 728.84 CAD
CA80056-570
L(-)-Glutathione (oxidised form) ≥95% (by HPLC)
L(-)-Glutathione (oxidized form)

Cyclic(γ-Glu-Cys-Gly)₂, GSSG


Glutathione, oxidized forms a 'redox buffer' in physiological systems in the presence of reduced glutathione.

Formula: C₂₀H₃₂N₆O₁₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00063106
CAS Number: 27025-41-8

Order Now

Specification Test Results

Form White Solid
Purity ≥95% by HPLC
Toxicity Irritant
Storage Temperature +2 °C to +8 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR