Estradiol 97, Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: beta-Estradiol, 17β-Estradiol

MQ100

3301-1GM
CA80056-424EA 232.26 CAD
CA80056-424
Estradiol 97, Calbiochem®, Sigma-Aldrich®
Estradiol

Most potent mammalian estrogenic hormone


Induces nitric oxide synthase in the aorta.

Formula: C₁₈H₂₄O₂
MW: 272.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR