Anti-1A4 Mouse Monoclonal Antibody [clone: 1A4]

Supplier: MilliporeSigma
113200-500UL
CA80055-622EA 1156.21 CAD
CA80055-622
Anti-1A4 Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
Primary Mouse Anti-alpha-Actin (1A4) Reacts with Baboon, Human, Rabbit (Lapine)

Type: Primary
Antigen: 1A4
Clonality: Monoclonal
Clone: 1A4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Baboon, Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR