Sodium hydroxide, pellets

Supplier: MilliporeSigma

Synonyms: Caustic soda

567530-250GM 567530-500GM
CA80055-562EA 234.83 CAD
CA80055-562 CA80055-564
Sodium hydroxide, pellets
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
MDL Number: MFCD00003548
CAS Number: 1310-73-2
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR