Glutaraldehyde 25% in aqueous solution

Supplier: MilliporeSigma

354400-500ML
CA80055-556EA 608.15 CAD
CA80055-556
Glutaraldehyde 25% in aqueous solution
Glutaraldehyde
Formula: C₅H₈O₂
MDL Number: MFCD00007025
CAS Number: 111-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR