Pig Pepsin (from Stomach mucosa)

Supplier: MilliporeSigma

516360-500MG
CA80053-620EA 167.76 CAD
CA80053-620
Pig Pepsin (from Stomach mucosa)
Enzymes

Pepsin is isolated from procine stomach mucosa. Catalyzes the hydrolysis of aminoacyl-proline to an amino acid and proline. Inhibitors include aliphatic alcohols, pepstatin A and pH >6.0.


  • Lyophilized form
  • Store at —20 °C


Unit definition: One unit is defined as the amount of enzyme that will digest hemoglobin substrates to TCA-soluble products, measured as a change in absorbance at 280 nm of 0.001 per min at 37 °C, pH 2.0.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR