Anti-MMP9 Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
444236-500UL
CA80053-088EA 1050.4 CAD
CA80053-088
Anti-MMP9 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-MMP-9 Reacts with Human, Marmoset

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Marmoset
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR