Creatine phosphate dipotassium salt ≥98% (by TLC), Calbiochem®

Supplier: MilliporeSigma

237911-250MG
CA80051-256EA 441.11 CAD
CA80051-256
Creatine phosphate dipotassium salt ≥98% (by TLC), Calbiochem®
Creatine phosphate dipotassium salt

Creatine phosphate, dipotassium salt is a high-energy phosphate compound for transfer of phosphate groups in the creatine phosphate energy shuttle.


Synonym: Phosphocreatine.

Formula: C₄H₈K₂N₃O₅P
Storage Temperature: Freezer
CAS Number: 18838-38-5
Merck Index: 14,07340

Order Now

Specification Test Results

Purity (TLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR