Anti-ALB Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
126705-1ML
CA80050-202EA 697.76 CAD
CA80050-202
Anti-ALB Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Albumin Reacts with Chicken

Type: Primary
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR