Anti-MSH2 Mouse Monoclonal Antibody

Supplier: MilliporeSigma
NA27-100UG
CA80000-852EA 1180.5 CAD
CA80000-852
Anti-MSH2 Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-MSH2 (Ab-2) (FE11) Reacts with Human, Mouse

Type: Primary
Antigen: MSH2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR