Anti-MMP9 Mouse Monoclonal Antibody

Supplier: MilliporeSigma
IM09L-100UG
CA80000-620EA 1583.47 CAD
CA80000-620
Anti-MMP9 Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-MMP-9 (Ab-1) (6-6B) Reacts with Human

Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR