Eucalyptol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.43788.0100 8.43788.0500 8.43788.0100 8.43788.0500
EM8.43788.0100CS 1038.06 CAD
EM8.43788.0100 EM8.43788.0500 CA8.43788.0100 CA8.43788.0500
Eucalyptol for synthesis, Sigma-Aldrich®
Eucalyptol
Formula: C₁₀H₁₈O
Boiling Pt: 176…177 °C (1013 hPa)
Melting Pt: 1.5 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00167977
CAS Number: 470-82-6
EINECS: 207-431-5
UN: 1993
ADR: 3,II
Merck Index: 12,03940

Order Now

Specification Test Results

Solubility 3.5 g/l (21 °C)
Melting Point 1.5 °C
Molar Mass 154.25 g/mol
Boiling Point 174 - 177 °C
Flash Point 49 °C
Density 0.925 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR