N,N-Dimethyloctylamine, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N,N-Dimethyloctan-1-amine

8.41863.0002
CA8.41863.0002EA 43 CAD
CA8.41863.0002
N,N-Dimethyloctylamine, Sigma-Aldrich®
N,N-Dimethyloctylamine
Formula: C₁₀H₂₃N
MW: 157.3 g/mol
Boiling Pt: 195
Melting Pt: -57
Density: 0.765 g/cm³ (20 °C)
Flash Pt: 65
MDL Number: MFCD00009558
CAS Number: 7378-99-6
UN: 2922
ADR: 8 (6.1) II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR