2-Nitrobenzyl chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.41296.0010 8.41296.0010
EM8.41296.0010CS 656896.06 CAD
EM8.41296.0010 CA8.41296.0010
2-Nitrobenzyl chloride for synthesis, Sigma-Aldrich®
2-Nitrobenzyl chloride
Formula: C₇H₆ClNO₂
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 49…51 °C
Density: 1.5557 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007185
CAS Number: 612-23-7
EINECS: 210-300-5
UN: 3457
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 46 - 49 °C
Molar Mass 171.58 g/mol
Boiling Point 127 - 133 °C (13 hPa)
Flash Point 112 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR