2,2'-Dipyridyl disulfide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.41109.0005
CA8.41109.0005EA 187 CAD
CA8.41109.0005
2,2'-Dipyridyl disulfide for synthesis, Sigma-Aldrich®
2,2'-Dipyridyl disulphide
Formula: C₁₀H₈N₂S₂
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 57…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006287
CAS Number: 2127-03-9
EINECS: 218-343-1

Order Now

Specification Test Results

Solubility 5 g/l (20 °C)
Melting Point 57 - 61 °C
Molar Mass 220.3 g/mol
Flash Point 210 °C
Ignition Point 340 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR