trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,2',2'',2'''-(trans-Cyclohexane-1,2-diylbis(azanetriyl))tetraacetic acid hydrate, DCTA monohydrate, CDTA monohydrate, trans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrate, (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate, 2,2',2'',2'''-[trans-Cyclohexane-1,2-diylbis(azanetriyl)]tetraacetic acid hydrate, CHELATON 4, Titriplex® IV, trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, trans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydrate

MQ200

8.41032.0025 8.41032.0100 8.41032.0025 8.41032.0100 8.41032.0005
EM8.41032.0025CS 13153.5 CAD
EM8.41032.0025 EM8.41032.0100 CA8.41032.0025 CA8.41032.0100 CA8.41032.0005
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate zS, Sigma-Aldrich®
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Formula: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 213 °C
Molar Mass 364.34 g/mol
Bulk Density 500 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR