Tetradecyltrimethylammonium bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB

M-Clarity™ quality level = MQ200

8.40006.0025 8.40006.0100 8.40006.0025 8.40006.0100
EM8.40006.0025CS 4493.52 CAD
EM8.40006.0025 EM8.40006.0100 CA8.40006.0025 CA8.40006.0100
Tetradecyltrimethylammonium bromide for synthesis, Sigma-Aldrich®
Tetradecyltrimethylammonium bromide
Formula: C₁₇H₃₈BrN
MW: 336.394 g/mol
Melting Pt: 245…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011770
CAS Number: 1119-97-7
EINECS: 214-291-9
UN: 3077
ADR: 9 III
Merck Index: 12,06419

Order Now

Specification Test Results

Solubility 100 g/l (20 °C)
Melting Point 245 - 250 °C
Molar Mass 336.4 g/mol
Bulk Density 600 kg/m3

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR