Mercaptoacetic acid (thioglycolic acid) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.22336.0250 8.22336.1000 8.22336.0250 8.22336.1000
EM8.22336.0250CS 676.24 CAD
EM8.22336.0250 EM8.22336.1000 CA8.22336.0250 CA8.22336.1000
Mercaptoacetic acid (thioglycolic acid) for synthesis, Sigma-Aldrich®
Mercaptoacetic acid (Thioglycolic acid)
Formula: HSCH₂COOH
Boiling Pt: 96 °C (1013 hPa)
Melting Pt: –16.5 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 130 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: 8,II
Merck Index: 13,09410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45179825145ec44f2e058197a0ee54845ae7c750&vid=51e43f5b69b880b196570b3065f9b4d8db7336d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -16.5 °C
Molar Mass 92.11 g/mol
Boiling Point 220 °C (1013 hPa) (decomposition)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 131.5 °C
Density 1.325 g/cm3 (20 °C)
pH 1 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR